Assay Details
| Gene Symbol: PRMT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099051.1 | NP_001092521.1 | 1 - 2 | 390 | 58 | |
| GenBank mRNA | BC142231.1 | - | 1 - 2 | 390 | 58 | |
More Information
Set Membership: |
|

PRMT9
Genomic Map