Assay Details
| Gene Symbol: MAP3K8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099071.2 | NP_001092541.1 | 2 - 3 | 445 | 74 | |
| GenBank mRNA | BC142398.1 | - | 2 - 3 | 446 | 74 | |
More Information
Set Membership: |
|

MAP3K8
Genomic Map