Assay Details
| Gene Symbol: ISL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099130.1 | NP_001092600.1 | 5 - 6 | 1155 | 56 | |
| GenBank mRNA | BC146163.1 | - | 5 - 6 | 1155 | 56 | |
More Information
Set Membership: |
|

ISL1
Genomic Map