Assay Details
| Gene Symbol: METTL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099135.2 | NP_001092605.1 | 1 - 2 | 419 | 117 | |
| GenBank mRNA | BC142084.1 | - | 1 - 2 | 420 | 117 | |
More Information
Set Membership: |
|

METTL5
Genomic Map