Assay Details
| Gene Symbol: SRD5A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099137.1 | NP_001092607.1 | 1 - 2 | 353 | 61 | |
| GenBank mRNA | BC142219.1 | - | 1 - 2 | 353 | 61 | |
More Information
Set Membership: |
|

SRD5A1
Genomic Map