Assay Details
| Gene Symbol: CYB561A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099149.1 | NP_001092619.1 | 2 - 3 | 243 | 62 | |
| GenBank mRNA | BC146220.1 | - | 2 - 3 | 243 | 62 | |
| BT030526.1 | - | 2 - 3 | 245 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC146220.1, NM_001099149.1 transcripts.
Set Membership: |
|

CYB561A3
Genomic Map