Assay Details
| Gene Symbol: TMEM41B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099156.1 | NP_001092626.1 | 4 - 5 | 640 | 80 | |
| GenBank mRNA | BC142213.1 | - | 4 - 5 | 640 | 80 | |
More Information
Set Membership: |
|

TMEM41B
Genomic Map