Assay Details
| Gene Symbol: BCL7C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099722.2 | NP_001093192.1 | 5 - 6 | 645 | 68 | |
| GenBank mRNA | BC142100.1 | - | 5 - 6 | 646 | 68 | |
More Information
Set Membership: |
|

BCL7C
Genomic Map