Assay Details
| Gene Symbol: DHX32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101144.1 | NP_001094614.1 | 8 - 9 | 1998 | 68 | |
| GenBank mRNA | BC151578.1 | - | 8 - 9 | 1998 | 68 | |
More Information
Set Membership: |
|

DHX32
Genomic Map