Assay Details
| Gene Symbol: CCL28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101163.1 | NP_001094633.1 | 2 - 3 | 221 | 89 | |
| GenBank mRNA | EF654535.1 | - | 2 - 3 | 221 | 89 | |
More Information
Set Membership: |
|

CCL28
Genomic Map