Assay Details
| Gene Symbol: SLC27A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101169.1 | NP_001094639.1 | 3 - 4 | 1041 | 73 | |
| GenBank mRNA | BC149774.1 | - | 3 - 4 | 1041 | 73 | |
| KP735933.1 | - | 3 - 4 | 715 | 73 | ||
More Information
Set Membership: |
|

SLC27A6
Genomic Map