Assay Details
| Gene Symbol: ISOC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101213.2 | NP_001094683.1 | 2 - 3 | 495 | 120 | |
| GenBank mRNA | BC148130.1 | - | 2 - 3 | 496 | 120 | |
More Information
Set Membership: |
|

ISOC1
Genomic Map