Assay Details
| Gene Symbol: CCDC92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101243.1 | NP_001094713.1 | 3 - 4 | 216 | 71 | |
| GenBank mRNA | BC149223.1 | - | 3 - 4 | 216 | 71 | |
More Information
Set Membership: |
|

CCDC92
Genomic Map