Assay Details
| Gene Symbol: DDX20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102118.1 | NP_001095588.1 | 8 - 9 | 1113 | 68 | |
| GenBank mRNA | BC151337.1 | - | 8 - 9 | 1113 | 68 | |
More Information
Set Membership: |
|

DDX20
Genomic Map