Assay Details
| Gene Symbol: TAPBPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102121.2 | NP_001095591.1 | 1 - 2 | 475 | 61 | |
| GenBank mRNA | BC148877.1 | - | 1 - 2 | 476 | 61 | |
More Information
Set Membership: |
|

TAPBPL
Genomic Map