Assay Details
| Gene Symbol: RNF145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102168.2 | NP_001095638.1 | 6 - 7 | 957 | 73 | |
| GenBank mRNA | BC148080.1 | - | 6 - 7 | 958 | 73 | |
More Information
Set Membership: |
|

RNF145
Genomic Map