Assay Details
| Gene Symbol: OTULINL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102171.1 | NP_001095641.1 | 3 - 4 | 262 | 95 | |
| GenBank mRNA | BC148069.1 | - | 3 - 4 | 262 | 95 | |
More Information
Set Membership: |
|

OTULINL
Genomic Map