Assay Details
| Gene Symbol: GRAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102173.1 | NP_001095643.1 | 2 - 3 | 250 | 140 | |
| GenBank mRNA | BC147998.1 | - | 2 - 3 | 250 | 140 | |
More Information
Set Membership: |
|

GRAP
Genomic Map