Assay Details
| Gene Symbol: MAPK14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102174.1 | NP_001095644.1 | 1 - 2 | 516 | 63 | |
| GenBank mRNA | BC148063.1 | - | 1 - 2 | 516 | 63 | |
More Information
Set Membership: |
|

MAPK14
Genomic Map