Assay Details
| Gene Symbol: FKBP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102209.1 | NP_001095679.1 | 3 - 4 | 289 | 56 | |
| GenBank mRNA | BC149679.1 | - | 3 - 4 | 289 | 56 | |
More Information
Set Membership: |
|

FKBP6
Genomic Map