Assay Details
| Gene Symbol: LRRC47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102298.1 | NP_001095768.1 | 1 - 2 | 625 | 71 | |
| GenBank mRNA | BC150059.1 | - | 1 - 2 | 625 | 71 | |
More Information
Set Membership: |
|

LRRC47
Genomic Map