Assay Details
| Gene Symbol: MCUR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102309.2 | NP_001095779.1 | 2 - 3 | 595 | 65 | |
| GenBank mRNA | BC149934.1 | - | 2 - 3 | 596 | 65 | |
More Information
Set Membership: |
|

MCUR1
Genomic Map