Assay Details
| Gene Symbol: DEPTOR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102312.1 | NP_001095782.1 | 5 - 6 | 827 | 58 | |
| GenBank mRNA | BC149596.1 | - | 5 - 6 | 827 | 58 | |
More Information
Set Membership: |
|

DEPTOR
Genomic Map