Assay Details
| Gene Symbol: HRC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102313.1 | NP_001095783.1 | 3 - 4 | 2628 | 60 | |
| GenBank mRNA | BC149780.1 | - | 3 - 4 | 2628 | 60 | |
More Information
Set Membership: |
|

HRC
Genomic Map