Assay Details
| Gene Symbol: RPAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103266.1 | NP_001096736.1 | 9 - 10 | 1560 | 80 | |
| GenBank mRNA | BC123867.1 | - | 9 - 10 | 1560 | 80 | |
More Information
Set Membership: |
|

RPAP2
Genomic Map