Assay Details
| Gene Symbol: TUBAL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103285.2 | NP_001096755.1 | 2 - 3 | 1129 | 71 | |
| GenBank mRNA | BC149967.1 | - | 2 - 3 | 1130 | 71 | |
More Information
Set Membership: |
|

TUBAL3
Genomic Map