Assay Details
| Gene Symbol: MED20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103293.1 | NP_001096763.1 | 3 - 4 | 499 | 82 | |
| GenBank mRNA | BC146258.1 | - | 3 - 4 | 499 | 82 | |
More Information
Set Membership: |
|

MED20
Genomic Map