Assay Details
| Gene Symbol: LMNB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103295.1 | NP_001096765.1 | 5 - 6 | 1306 | 71 | |
| GenBank mRNA | BC151304.1 | - | 5 - 6 | 1306 | 71 | |
More Information
Set Membership: |
|

LMNB1
Genomic Map