Assay Details
| Gene Symbol: FAM32A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105004.1 | NP_001098474.1 | 3 - 4 | 299 | 63 | |
| GenBank mRNA | BC150092.1 | - | 3 - 4 | 299 | 63 | |
More Information
Set Membership: |
|

FAM32A
Genomic Map