Assay Details
| Gene Symbol: TRIM17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105011.1 | NP_001098481.1 | 4 - 5 | 854 | 64 | |
| GenBank mRNA | BC111201.1 | - | 4 - 5 | 854 | 64 | |
More Information
Set Membership: |
|

TRIM17
Genomic Map