Assay Details
| Gene Symbol: RHBDF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105400.1 | NP_001098870.1 | 13 - 14 | 1769 | 58 | |
| GenBank mRNA | BC134588.1 | - | 13 - 14 | 1769 | 58 | |
More Information
Set Membership: |
|

RHBDF1
Genomic Map