Assay Details
| Gene Symbol: EFNA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105429.1 | NP_001098899.1 | 4 - 5 | 613 | 114 | |
| GenBank mRNA | BC134650.1 | - | 4 - 5 | 613 | 114 | |
More Information
Set Membership: |
|

EFNA3
Genomic Map