Assay Details
| Gene Symbol: UGT1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105636.1 | NP_001099106.1 | 1 - 2 | 884 | 73 | |
| GenBank mRNA | BC134515.1 | - | 1 - 2 | 884 | 73 | |
More Information
Set Membership: |
|

UGT1A1
Genomic Map