Assay Details
| Gene Symbol: SYNC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109803.2 | NP_001103273.1 | 3 - 4 | 1454 | 90 | |
| GenBank mRNA | BC149013.1 | - | 3 - 4 | 1455 | 90 | |
More Information
Set Membership: |
|

SYNC
Genomic Map