Assay Details
| Gene Symbol: ATP5MD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113318.1 | NP_001106789.1 | 1 | 243 | 131 | |
| GenBank mRNA | BC103275.1 | - | 1 | 243 | 131 | |
More Information
Set Membership: |
|

ATP5MD
Genomic Map