Assay Details
| Gene Symbol: UBE2U | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099193.1 | NP_001092663.1 | 3 - 4 | 673 | 76 | |
| GenBank mRNA | BC146108.1 | - | 3 - 4 | 673 | 76 | |
More Information
Set Membership: |
|

UBE2U
Genomic Map