Assay Details
| Gene Symbol: ATXN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099216.2 | NP_001092686.1 | 8 - 9 | 783 | 72 | |
| GenBank mRNA | BC146166.1 | - | 8 - 9 | 784 | 72 | |
More Information
Set Membership: |
|

ATXN3
Genomic Map