Assay Details
| Gene Symbol: CMTM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099399.1 | NP_001092869.1 | 4 - 5 | 562 | 63 | |
| GenBank mRNA | BC140678.1 | - | 4 - 5 | 562 | 63 | |
More Information
Set Membership: |
|

CMTM3
Genomic Map