Assay Details
| Gene Symbol: ABHD17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101278.1 | NP_001094748.1 | 2 - 3 | 815 | 82 | |
| GenBank mRNA | BC151543.1 | - | 2 - 3 | 815 | 82 | |
More Information
Set Membership: |
|

ABHD17B
Genomic Map