Assay Details
| Gene Symbol: NPY5R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102527.2 | NP_001095997.1 | 1 - 2 | 26 | 81 | |
| GenBank mRNA | BC150033.1 | - | 1 - 2 | 27 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC150033.1, NM_001102527.2 transcripts.
Set Membership: |
|

NPY5R
Genomic Map