Assay Details
| Gene Symbol: PRICKLE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102534.1 | NP_001096004.1 | 6 - 7 | 909 | 81 | |
| GenBank mRNA | BC140671.1 | - | 6 - 7 | 909 | 81 | |
More Information
Set Membership: |
|

PRICKLE1
Genomic Map