Assay Details
| Gene Symbol: CDC42SE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102537.1 | NP_001096007.1 | 4 - 5 | 686 | 95 | |
| GenBank mRNA | BC147986.1 | - | 4 - 5 | 686 | 95 | |
More Information
Set Membership: |
|

CDC42SE2
Genomic Map