Assay Details
| Gene Symbol: CDC42SE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102537.1 | NP_001096007.1 | 5 - 6 | 790 | 64 | |
| GenBank mRNA | BC147986.1 | - | 5 - 6 | 790 | 64 | |
More Information
Set Membership: |
|

CDC42SE2
Genomic Map