Assay Details
| Gene Symbol: GNAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102554.2 | NP_001096024.1 | 6 - 7 | 664 | 65 | |
| GenBank mRNA | BC149402.1 | - | 6 - 7 | 665 | 65 | |
More Information
Set Membership: |
|

GNAL
Genomic Map