Assay Details
| Gene Symbol: SELENOT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103103.2 | NP_001096573.2 | 1 - 2 | 253 | 88 | |
| GenBank mRNA | BC149082.1 | - | 1 - 2 | 247 | 88 | |
| DT887648.1 | - | 3 - 4 | 297 | 88 | ||
| DV816938.1 | - | 1 - 2 | 184 | 88 | ||
| DY197381.1 | - | 1 - 2 | 254 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC149082.1 transcripts.
Set Membership: |
|

SELENOT
Genomic Map