Assay Details
| Gene Symbol: SELENOT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103103.2 | NP_001096573.2 | 4 - 5 | 580 | 79 | |
| NR_146805.1 | 5 - 6 | 679 | 79 | |||
| GenBank mRNA | BC149082.1 | - | 4 - 5 | 574 | 79 | |
| BF773952.1 | - | 1 - 2 | 79 | 79 | ||
| DV816938.1 | - | 4 - 5 | 511 | 79 | ||
| DV833984.1 | - | 5 - 6 | 581 | 79 | ||
More Information
Set Membership: |
|

SELENOT
Genomic Map