Assay Details
| Gene Symbol: B4GALT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105030.2 | NP_001098500.1 | 6 - 7 | 1175 | 66 | |
| GenBank mRNA | BC150042.1 | - | 6 - 7 | 1176 | 66 | |
More Information
Set Membership: |
|

B4GALT2
Genomic Map