Assay Details
| Gene Symbol: TLCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105042.1 | NP_001098512.1 | 1 - 2 | 692 | 78 | |
| GenBank mRNA | BC140551.1 | - | 1 - 2 | 692 | 78 | |
More Information
Set Membership: |
|

TLCD1
Genomic Map