Assay Details
| Gene Symbol: SNTB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105466.1 | NP_001098936.1 | 2 - 3 | 815 | 89 | |
| GenBank mRNA | BC134712.1 | - | 2 - 3 | 815 | 89 | |
More Information
Set Membership: |
|

SNTB2
Genomic Map