Assay Details
| Gene Symbol: BOLA-DQB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012676.3 | NP_001012694.2 | 2 - 3 | 455 | 150 | |
| GenBank mRNA | AF037315.1 | - | 2 - 3 | 460 | 150 | |
| BC133449.1 | - | 2 - 3 | 456 | 150 | ||
More Information
Set Membership: |
|

BOLA-DQB
Genomic Map